2-Bromopyridine, 99%
2-Bromopyridine, 99%
2-Bromopyridine, 99%
Thermo Scientific Chemicals

2-Bromopyridine, 99%

CAS: 109-04-6 | C5H4BrN | 157.998 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
500 g
Catalog number A13241.22
also known as A13241-22
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS109-04-6
IUPAC Name2-bromopyridine
Molecular FormulaC5H4BrN
InChI KeyIMRWILPUOVGIMU-UHFFFAOYSA-N
SMILESBrC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5695-1.5725 @ 20?C
Appearance (Color)Clear colorless to pale yellow or pale brown
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

Solubility
Slightly miscible with water.

Notes
Keep away from heat, sparks, and flame. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and acid chlorides.
RUO – Research Use Only

General References:

  1. Bromopyridines can be converted to the Grignards by Mg exchange with i-PrMgCl: Tetrahdron Lett., 40, 4339 (1999).
  2. Reaction with n-BuLi gives 2-lithio-pyridine. Subsequent reaction at low temperatures with PCl3 gives, as the major product, 2,2'-bipyridine, along with a low yield of the expected tris(2-pyridyl)phosphine. POCl3 or SOCl2 also promote the symmetrical coupling reaction: Heteroatom. Chem., 5, 409 (1994). In contrast, reaction with LDA in THF at -78° results in lithiation at the 3-position, giving access to 3-substituted 2-bromopyridines: Synthesis, 235, 237 (1982). See also: Org. Lett., 3, 835 (2001).
  3. Kathiravan, S.; Ghosh, S.; Hogarth, G.; Nicholls, I. A. Copper catalysed amidation of aryl halides through chelation assistance. Chem. Commun. 2015, 51 (23), 4834-4837.
  4. Liu, C.; Li, X.; Liu, C.; Wang, X.; Qiu, J. Palladium-catalyzed ligand-free and efficient Suzuki-Miyaura reaction of heteroaryl halides with MIDA boronates in water. RSC Adv. 2015, 5 (67), 54312-54315.