2,6-Dichlorobenzaldehyde, 97+%
2,6-Dichlorobenzaldehyde, 97+%
2,6-Dichlorobenzaldehyde, 97+%
Thermo Scientific Chemicals

2,6-Dichlorobenzaldehyde, 97+%

CAS: 83-38-5 | C7H4Cl2O | 175.008 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13295.22
also known as A13295-22
Price (USD)
84.65
Online Exclusive
94.00
Save 9.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
94.00
Save 9.35 (10%)
Each
Chemical Identifiers
CAS83-38-5
IUPAC Name2,6-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyDMIYKWPEFRFTPY-UHFFFAOYSA-N
SMILESClC1=CC=CC(Cl)=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormCrystals or powder or crystalline powder or lumps or flakes
Assay (GC)≥97.0%
Free acid (titration)≤3%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only