L-Citrulline, 98%
L-Citrulline, 98%
L-Citrulline, 98%
Thermo Scientific Chemicals

L-Citrulline, 98%

CAS: 372-75-8 | C6H13N3O3 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13316.18
also known as A13316-18
Price (USD)
183.65
Online Exclusive
204.00
Save 20.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
183.65
Online Exclusive
204.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS372-75-8
IUPAC Name2-amino-5-(carbamoylamino)pentanoic acid
Molecular FormulaC6H13N3O3
InChI KeyRHGKLRLOHDJJDR-UHFFFAOYNA-N
SMILESNC(CCCNC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation22.5 ± 1.0? (C=2 in 1N HCl)
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.

Solubility
It is soluble in water.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
RUO – Research Use Only

General References:

  1. F Cosentin.; S Patton.; LV d'Uscio. Tetrahydrobiopterin alters superoxide and nitric oxide release in prehypertensive rats. J Clin Invest. .1998, 101,(7), 1530-1537.
  2. Y Xia.; LJ Roman.; BSS Masters.; JL Zweier. Inducible nitric-oxide synthase generates superoxide from the reductase domain. Journal of Biological Chemistry.1998, 273,(35), 22635-22639.