3-Bromo-4-fluorobenzonitrile, 98%
3-Bromo-4-fluorobenzonitrile, 98%
3-Bromo-4-fluorobenzonitrile, 98%
3-Bromo-4-fluorobenzonitrile, 98%
Thermo Scientific Chemicals

3-Bromo-4-fluorobenzonitrile, 98%

CAS: 79630-23-2 | C7H3BrFN | 200.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13318.14
also known as A13318-14
Price (USD)
85.65
Online Exclusive
95.20
Save 9.55 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.20
Save 9.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS79630-23-2
IUPAC Name3-bromo-4-fluorobenzonitrile
Molecular FormulaC7H3BrFN
InChI KeyJKCYKISVUIVZCS-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1Br)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Melting Point (clear melt)52.0-59.0?C
Appearance (Color)White
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only