1-Bromo-4-(n-octyloxy)benzene, 98%
1-Bromo-4-(n-octyloxy)benzene, 98%
1-Bromo-4-(n-octyloxy)benzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-(n-octyloxy)benzene, 98%

CAS: 96693-05-9 | C14H21BrO | 285.225 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number A13334.14
also known as A13334-14
Price (USD)
364.65
Online Exclusive
405.00
Save 40.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
364.65
Online Exclusive
405.00
Save 40.35 (10%)
Each
Chemical Identifiers
CAS96693-05-9
IUPAC Name1-bromo-4-(octyloxy)benzene
Molecular FormulaC14H21BrO
InChI KeyUVBFFPZGOOKWNR-UHFFFAOYSA-N
SMILESCCCCCCCCOC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5155-1.5195 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only