2-Hexyn-1-ol, 97%
2-Hexyn-1-ol, 97%
2-Hexyn-1-ol, 97%
Thermo Scientific Chemicals

2-Hexyn-1-ol, 97%

CAS: 764-60-3 | C6H10O | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number A13339.06
also known as A13339-06
Price (USD)
68.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
68.90
Each
Chemical Identifiers
CAS764-60-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.4520-1.4570 @ 20?C
Assay (GC)≥96.0%
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Avoid temperature and direct sunlight.
RUO – Research Use Only

General References:

  1. W Zhang.; DP Curran.; CHT Chen. Use of fluorous silica gel to separate fluorous thiol quenching derivatives in solution-phase parallel synthesis. Tetrahedron letters. 2004, 45,(29), 5573-5575.
  2. DF Taber.; RJ Herr.; DM Gleave. Diastereoselective synthesis of an isoprostane:(±)-8-epi-PGF2α ethyl ester. J. Org. Chem. 1997, 62,(1), 194-198.