Tri-n-butyltin methoxide, 97%
Tri-n-butyltin methoxide, 97%
Tri-n-butyltin methoxide, 97%
Thermo Scientific Chemicals

Tri-n-butyltin methoxide, 97%

CAS: 1067-52-3 | C13H30OSn | 321.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A13355.0B
also known as A13355-0B
Price (USD)
-
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS1067-52-3
IUPAC Name1,1,1-tributyl-O-methylstannanol
Molecular FormulaC13H32OSn
InChI KeySIIBSBQSPWMULI-UHFFFAOYSA-N
SMILESCO.CCCC[SnH](CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisSn: 35.5-38.45% (Theory: 36.97%)
Appearance (Color)Clear colorless
Elemental AnalysisH: 9.03-9.79% (Theory: 9.41%)
FormLiquid
Elemental AnalysisC: 46.69-50.58% (Theory: 48.63%)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only