4-n-Hexyloxybenzoyl chloride, 98%
4-n-Hexyloxybenzoyl chloride, 98%
4-n-Hexyloxybenzoyl chloride, 98%
Thermo Scientific Chemicals

4-n-Hexyloxybenzoyl chloride, 98%

CAS: 39649-71-3 | C13H17ClO2 | 240.727 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number A13367.06
also known as A13367-06
Price (USD)
65.65
Online Exclusive
73.10
Save 7.45 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.65
Online Exclusive
73.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS39649-71-3
IUPAC Name4-(hexyloxy)benzoyl chloride
Molecular FormulaC13H17ClO2
InChI KeyDQQOONVCLQZWOY-UHFFFAOYSA-N
SMILESCCCCCCOC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5355-1.5395 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only