1-Bromo-4-isopropylbenzene, 97%
1-Bromo-4-isopropylbenzene, 97%
1-Bromo-4-isopropylbenzene, 97%
Thermo Scientific Chemicals

1-Bromo-4-isopropylbenzene, 97%

CAS: 586-61-8 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number A13370.09
also known as A13370-09
Price (USD)
126.65
Online Exclusive
141.00
Save 14.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
141.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS586-61-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.5340-1.5390 @ 20?C
FormLiquid
It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Insoluble in water

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Michael C. Varela; Seth M. Dixon; Michael D. Price; Jeffrey E. Merit; Patrick E. Berget; Saori Shiraki; Mark J. Kurth; Neil E. Schore. Comparison of enantioselective reductions using bead and monolith ‘disk’ polymer formulations of CBS catalysts. Tetrahedron,. 2007, 63 (16), 3334-3339.
  2. Masato Ofuji; Andrew J. Lovinger; Christian Kloc; Theo Siegrist; Ashok J. Maliakal; and Howard E. Katz. Organic Semiconductor Designed for Lamination Transfer between Polymer Films. Chem. Mater, 2005, 17 (23), 5748-5753.