2,3-Dichloro-1,4-naphthoquinone, 98%
2,3-Dichloro-1,4-naphthoquinone, 98%
2,3-Dichloro-1,4-naphthoquinone, 98%
Thermo Scientific Chemicals

2,3-Dichloro-1,4-naphthoquinone, 98%

CAS: 117-80-6 | C10H4Cl2O2 | 227.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13378.22
also known as A13378-22
Price (USD)
53.65
Online Exclusive
59.80
Save 6.15 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS117-80-6
IUPAC Name2,3-dichloro-1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC10H4Cl2O2
InChI KeySVPKNMBRVBMTLB-UHFFFAOYSA-N
SMILESClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)191.0-198.0°C
Use in textiles, as organic catalyst, as chemical intermediate, as additive in dye binders. Also used as an important catalyst to produce 3,3-dichlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as seed disinfectant, textiles, as organic catalyst, as chemical intermediate, as additive in dye binders, as antifouling agent. Also used to control blue-green algae in ponds, lakes, and swimming pools. Also used as an important catalyst to produce 3,3-Dichloro benzine.

Solubility
Soluble in water (0.008g/L). Soluble in hot toluene almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. R M Vos; B Van Ommen; M S Hoekstein; J H De Goede; P J Van Bladeren. Irreversible inhibition of rat hepatic glutathione S-transferase isoenzymes by a series of structurally related quinones. Chemico-Biological Interactions. 1989, 71 (4), 381-392.
  2. N Gallo; P Bianco; R Tapino; G Luisi. Interaction of methimazole and quinones. Spectrophotometric determination of methimazole in blood. Minerva medica, 1983, 74 (16), 875-877.
  3. Reagent for the spectrophotometric determination of hydrazides: Anal. Chem., 48, 1536 (1976).