2-Mercaptopyrimidine, 98%
2-Mercaptopyrimidine, 98%
2-Mercaptopyrimidine, 98%
2-Mercaptopyrimidine, 98%
Thermo Scientific Chemicals

2-Mercaptopyrimidine, 98%

CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
250 g
Catalog number A13382.30
also known as A13382-30
Price (USD)
673.65
Online Exclusive
748.00
Save 74.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
673.65
Online Exclusive
748.00
Save 74.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1450-85-7
IUPAC Name1,2-dihydropyrimidine-2-thione
Molecular FormulaC4H4N2S
InChI KeyHBCQSNAFLVXVAY-UHFFFAOYSA-N
SMILESS=C1NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only