4-Fluoromandelic acid, 98%
4-Fluoromandelic acid, 98%
4-Fluoromandelic acid, 98%
4-Fluoromandelic acid, 98%
Thermo Scientific Chemicals

4-Fluoromandelic acid, 98%

CAS: 395-33-5 | C8H7FO3 | 170.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13391.03
also known as A13391-03
Price (USD)
18.60
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
18.60
Each
Add to cart
Chemical Identifiers
CAS395-33-5
IUPAC Name2-(4-fluorophenyl)-2-hydroxyacetic acid
Molecular FormulaC8H7FO3
InChI KeyRWCMOQXHIDWDDJ-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)134.0-140.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only