N,N'-Diacetylhydrazine, 98%
N,N'-Diacetylhydrazine, 98%
N,N'-Diacetylhydrazine, 98%
Thermo Scientific Chemicals

N,N'-Diacetylhydrazine, 98%

CAS: 3148-73-0 | C4H8N2O2 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13393.30
also known as A13393-30
Price (USD)
639.65
Online Exclusive
710.00
Save 70.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
639.65
Online Exclusive
710.00
Save 70.35 (10%)
Each
Chemical Identifiers
CAS3148-73-0
IUPAC NameN'-acetylacetohydrazide
Molecular FormulaC4H8N2O2
InChI KeyZLHNYIHIHQEHJQ-UHFFFAOYSA-N
SMILESCC(=O)NNC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)134-143°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only