2-Phenylethylamine, 99%
2-Phenylethylamine, 99%
2-Phenylethylamine, 99%
2-Phenylethylamine, 99%
Thermo Scientific Chemicals

2-Phenylethylamine, 99%

CAS: 64-04-0 | C8H11N | 121.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1000 mL
Catalog number A13402.AU
also known as A13402-AU
Price (USD)
214.00
Each
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (USD)
214.00
Each
Add to cart
Chemical Identifiers
CAS64-04-0
IUPAC Name2-phenylethan-1-amine
Molecular FormulaC8H11N
InChI KeyBHHGXPLMPWCGHP-UHFFFAOYSA-N
SMILESNCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Refractive Index1.5320-1.5350 @ 20°C
Assay (GC)≥98.5%
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Store under inert gas. Keep away from acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide (CO2).
RUO – Research Use Only

General References:

  1. HC Sabelli.; AD Mosnaim. Phenylethylamine hypothesis of affective behavior. American Journal of Psychiatry. 1974, 131,(6), 695-699.
  2. DA Durden.; SR Philips. Identification and distribution of β-phenylethylamine in the rat. Canadian Journal of Biochemistry. 1973, 51,(7), 995-1002.
  3. N-Acyl-derivatives are cyclized by a variety of reagents, e.g. P2O5, POCl3, PCl5, etc., to derivatives of isoquinoline, in the Bischler-Napieralski synthesis. For a review, see: Org. React., 6, 74 (1951). Similarly, imines can be cyclized to derivatives of tetrahydroisoquinoline in the Pictet-Spengler synthesis. Reviews: Org. React., 6, 151 (1951); Adv. Het. Chem., 3, 79 (1964); Heterocycles, 3, 223 (1975); 39, 903 (1994).