3-Chlorocinnamic acid, predominantly trans, 98+%
3-Chlorocinnamic acid, predominantly trans, 98+%
3-Chlorocinnamic acid, predominantly trans, 98+%
3-Chlorocinnamic acid, predominantly trans, 98+%
Thermo Scientific Chemicals

3-Chlorocinnamic acid, predominantly trans, 98+%

CAS: 1866-38-2 | C9H7ClO2 | 182.603 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13416.18
also known as A13416-18
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1866-38-2
IUPAC Name(2E)-3-(3-chlorophenyl)prop-2-enoic acid
Molecular FormulaC9H7ClO2
InChI KeyFFKGOJWPSXRALK-SNAWJCMRSA-N
SMILESOC(=O)C=CC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)159.0-166.0?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (HPLC)≥98.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only