Cyclobutylamine, 98%
Cyclobutylamine, 98%
Cyclobutylamine, 98%
Cyclobutylamine, 98%
Thermo Scientific Chemicals

Cyclobutylamine, 98%

CAS: 2516-34-9 | C4H9N | 71.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13423.03
also known as A13423-03
Price (USD)
53.65
Online Exclusive
59.10
Save 5.45 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.10
Save 5.45 (9%)
Each
Add to cart
Chemical Identifiers
CAS2516-34-9
IUPAC Namecyclobutanamine
Molecular FormulaC4H9N
InChI KeyKZZKOVLJUKWSKX-UHFFFAOYSA-N
SMILESNC1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4355-1.4395 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

Solubility
Slightly soluble in water.

Notes
Hygroscopic, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. McNaughton D; and Evans CJ J. The High-Resolution FTIR Far-Infrared Spectrum of Vinylamine. Mol. Spectrosc. 1999, 196 (2), 274-282,.
  2. Hartmut Schirok. Microwave-assisted flexible synthesis of 7-azaindoles. Journal of Organic Chemistry, 2006, 71 (15), 5538-5545.