1,4-Diphenylbutadiyne, 99%
1,4-Diphenylbutadiyne, 99%
1,4-Diphenylbutadiyne, 99%
Thermo Scientific Chemicals

1,4-Diphenylbutadiyne, 99%

CAS: 886-66-8 | C16H10 | 202.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13471.06
also known as A13471-06
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS886-66-8
IUPAC Name(4-phenylbuta-1,3-diyn-1-yl)benzene
Molecular FormulaC16H10
InChI KeyHMQFJYLWNWIYKQ-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C#CC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)84.0-91.0°C
1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

Solubility
Sparingly soluble in water(5.5E-4 g/L) (25°C). Soluble in toluene almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Norihito Fukui et. al. Synthesis of 7,8-dehydropurpurin dimers and their conversion into conformationally constrained β-to-β vinylene-bridged porphyrin dimers. Angewandte Chemie. International edition in English. 2014, 53(17), 4395-4398.
  2. Ajayi-Obe T, et al.1, 4-Diphenylbutadiyne complexes of tungsten (II). J. Organomet. Chem., 1994, 468 (1), 165-70.