2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
Thermo Scientific Chemicals

2,3-Dihydroxybenzaldehyde, 97%

CAS: 24677-78-9 | C7H6O3 | 138.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13490.06
also known as A13490-06
Price (USD)
53.65
Online Exclusive
59.70
Save 6.05 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS24677-78-9
IUPAC Name2,3-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow-green or cream to brown or grey
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Clarity5% w/v in methanol: clear solution
Melting Point (clear melt)103-109?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only