3,5-Bis(trifluoromethyl)benzhydrazide, 97%
3,5-Bis(trifluoromethyl)benzhydrazide, 97%
3,5-Bis(trifluoromethyl)benzhydrazide, 97%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzhydrazide, 97%

CAS: 26107-82-4 | C9H6F6N2O | 272.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13492.14
also known as A13492-14
Price (USD)
486.65
Online exclusive
541.00
Save 54.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
486.65
Online exclusive
541.00
Save 54.35
Each
Chemical Identifiers
CAS26107-82-4
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)128.0-138.0?C
Appearance (Color)White to pale brown
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only