2-Chloro-4-fluorothiophenol, 97%
2-Chloro-4-fluorothiophenol, 97%
2-Chloro-4-fluorothiophenol, 97%
Thermo Scientific Chemicals

2-Chloro-4-fluorothiophenol, 97%

CAS: 175277-99-3 | C6H4ClFS | 162.606 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13498.06
also known as A13498-06
Price (USD)
261.65
Online Exclusive
291.00
Save 29.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
261.65
Online Exclusive
291.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS175277-99-3
IUPAC Name(2-chloro-4-fluorophenyl)sulfanide
Molecular FormulaC6H3ClFS
InChI KeyKUAPPJSILOMQPC-UHFFFAOYSA-M
SMILESFC1=CC=C([S-])C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5650-1.5700 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only