2-Methoxybenzoic acid, 98+%
2-Methoxybenzoic acid, 98+%
2-Methoxybenzoic acid, 98+%
Thermo Scientific Chemicals

2-Methoxybenzoic acid, 98+%

CAS: 579-75-9 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A13513.22
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.90
Save 7.25
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.90
Save 7.25
Each
Chemical Identifiers
CAS579-75-9
IUPAC Name2-methoxybenzoic acid
Molecular FormulaC8H8O3
InChI KeyILUJQPXNXACGAN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)>98.0%
Appearance (Color)White to pale cream
Assay (Silylated GC)>98.0%
Formcrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only