1,4-Bis(2-methylstyryl)benzene, 99%
1,4-Bis(2-methylstyryl)benzene, 99%
1,4-Bis(2-methylstyryl)benzene, 99%
Thermo Scientific Chemicals

1,4-Bis(2-methylstyryl)benzene, 99%

CAS: 13280-61-0 | C24H22 | 310.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13534.06
also known as A13534-06
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.30
Save 11.65 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.30
Save 11.65 (15%)
Each
Chemical Identifiers
CAS13280-61-0
IUPAC Name1-methyl-2-[(1E)-2-{4-[(1E)-2-(2-methylphenyl)ethenyl]phenyl}ethenyl]benzene
Molecular FormulaC24H22
InChI KeyQKLPIYTUUFFRLV-YTEMWHBBSA-N
SMILESCC1=CC=CC=C1C=CC1=CC=C(C=CC2=CC=CC=C2C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or yellow-green or pale green to green
FormCrystals or crystalline powder or powder
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)176-185°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only