Thermo Scientific Chemicals

Cyclopropyl methyl ketone, 99%, Thermo Scientific Chemicals

Catalog number: A13540.22
100 g, Each
Thermo Scientific Chemicals

Cyclopropyl methyl ketone, 99%, Thermo Scientific Chemicals

Catalog number: A13540.22
100 g, Each
Quantity
Catalog number: A13540.22
also known as A13540-22
Price (USD)
Price: 126.00
Online price: 108.65
Your price:
Quantity
-

Chemical Identifiers

CAS
765-43-5
IUPAC Name
1-cyclopropylethan-1-one
Molecular Formula
C5H8O
InChI Key
HVCFCNAITDHQFX-UHFFFAOYSA-N
SMILES
CC(=O)C1CC1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4230-1.4260 @ 20?C

Description

Cyclopropyl methyl ketone was used in the preparation of homoallylic alcohols. Acetylcyclopropane is used as an intermediate for the manufacturing pharmaceuticals, agrochemicals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl methyl ketone was used in the preparation of homoallylic alcohols. Acetylcyclopropane is used as an intermediate for the manufacturing pharmaceuticals, agrochemicals and other organic compounds.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text