3,4-Dimethoxyaniline, 98+%
3,4-Dimethoxyaniline, 98+%
3,4-Dimethoxyaniline, 98+%
Thermo Scientific Chemicals

3,4-Dimethoxyaniline, 98+%

CAS: 6315-89-5 | C8H11NO2 | 153.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13548.18
also known as A13548-18
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS6315-89-5
IUPAC Name3,4-dimethoxyaniline
Molecular FormulaC8H11NO2
InChI KeyLGDHZCLREKIGKJ-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)84.5-90.5?C
Appearance (Color)Cream to brown
Assay (GC)≥98.0%
FormPowder
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

Solubility
Slightly soluble in water

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only