n-Heptyl 4-hydroxybenzoate, 98%
n-Heptyl 4-hydroxybenzoate, 98%
n-Heptyl 4-hydroxybenzoate, 98%
Thermo Scientific Chemicals

n-Heptyl 4-hydroxybenzoate, 98%

CAS: 1085-12-7 | C14H20O3 | 236.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13555.14
also known as A13555-14
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS1085-12-7
IUPAC Nameheptyl 4-hydroxybenzoate
Molecular FormulaC14H20O3
InChI KeyZTJORNVITHUQJA-UHFFFAOYSA-N
SMILESCCCCCCCOC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥97.5%
FormPowder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.

Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only