trans-2-Hexenoic acid, 96%
trans-2-Hexenoic acid, 96%
trans-2-Hexenoic acid, 96%
Thermo Scientific Chemicals

trans-2-Hexenoic acid, 96%

CAS: 13419-69-7 | C6H9O2 | 113.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13571.09
also known as A13571-09
Price (USD)
31.65
Online Exclusive
35.50
Save 3.85 (11%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
35.50
Save 3.85 (11%)
Each
Chemical Identifiers
CAS13419-69-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (Silylated GC)≥95.0%
Melting Point (clear melt)28.0-37.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only