Pentafluorobenzonitrile, 98%
Pentafluorobenzonitrile, 98%
Pentafluorobenzonitrile, 98%
Pentafluorobenzonitrile, 98%
Thermo Scientific Chemicals

Pentafluorobenzonitrile, 98%

CAS: 773-82-0 | C7F5N | 193.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13575.06
also known as A13575-06
Price (USD)
28.65
Online Exclusive
31.70
Save 3.05 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
28.65
Online Exclusive
31.70
Save 3.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS773-82-0
IUPAC Namepentafluorobenzonitrile
Molecular FormulaC7F5N
InChI KeyYXWJGZQOGXGSSC-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C#N)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4405-1.4445 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only