9-(Chloromethyl)anthracene, 98+%
9-(Chloromethyl)anthracene, 98+%
9-(Chloromethyl)anthracene, 98+%
Thermo Scientific Chemicals

9-(Chloromethyl)anthracene, 98+%

CAS: 24463-19-2 | C15H11Cl | 226.703 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13579.06
also known as A13579-06
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS24463-19-2
IUPAC Name9-(chloromethyl)anthracene
Molecular FormulaC15H11Cl
InChI KeyPCVRSXXPGXRVEZ-UHFFFAOYSA-N
SMILESClCC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Korte, W.D. 9(chloromethyl)anthracene: a useful derivatizing reagent for enhanced ultraviolet and fluorescence detection of carboxylic acids with liquid chromatography. Journal of Chromatography A. 1982, 243, (1), 153-157.
  2. Reagent for the protection of carboxylic acids, alcohols, phenols and thiols as their 9-anthrylmethyl derivatives, which are stable to mild acid or base, but specifically cleaved by NaSMe: J. Am. Chem. Soc., 96, 590 (1974). For use in N-protection, see: Aust. J. Chem., 18, 1699 (1965).