2,6-Difluorobenzenesulfonyl chloride, 97%
2,6-Difluorobenzenesulfonyl chloride, 97%
2,6-Difluorobenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

2,6-Difluorobenzenesulfonyl chloride, 97%

CAS: 60230-36-6 | C6H3ClF2O2S | 212.595 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13595.14
also known as A13595-14
Price (USD)
639.65
Special offer
Online exclusive
Ends: 30-Jun-2026
752.00
Save 112.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
639.65
Special offer
Online exclusive
Ends: 30-Jun-2026
752.00
Save 112.35 (15%)
Each
Chemical Identifiers
CAS60230-36-6
IUPAC Name2,6-difluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3ClF2O2S
InChI KeyQXWAUQMMMIMLTO-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥96.0%
Refractive Index1.5235-1.5285 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only