2-Methoxy-4-methylphenol, 98+%
2-Methoxy-4-methylphenol, 98+%
2-Methoxy-4-methylphenol, 98+%
Thermo Scientific Chemicals

2-Methoxy-4-methylphenol, 98+%

CAS: 93-51-6 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13603.09
also known as A13603-09
Price (USD)
27.65
Online Exclusive
30.10
Save 2.45 (8%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.10
Save 2.45 (8%)
Each
Chemical Identifiers
CAS93-51-6
IUPAC Name2-methoxy-4-methylphenol
Molecular FormulaC8H10O2
InChI KeyPETRWTHZSKVLRE-UHFFFAOYSA-N
SMILESCOC1=CC(C)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5350-1.5400 @ 20?C
Appearance (Color)Clear colorless to pale yellow
CommentMay turn yellow on storage
Assay (GC)≥98.0%
View more
2-Methoxy-4-methylphenol is used in preparation of renewable bis(cyanate) esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxy-4-methylphenol is used in preparation of renewable bis(cyanate) esters.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Grażyna Bieniek. Simultaneous determination of 2-methoxyphenol, 2-methoxy-4-methylphenol, 2,6-dimethoxyphenol and 4'-hydroxy-3'-methoxyacetophenone in urine by capillary gas chromatography. Journal of Chromatography B. 1993, 795 (2), 389-394.
  2. Heather A. Meylemans.; Benjamin G. Harvey.; Josiah T. Reams.; Andrew J. Guenthner.; Lee R. Cambrea.; Thomas J. Groshens.; Lawrence C. Baldwin.; Michael D. Garrison.; Joseph M. Mabry. Synthesis, Characterization, and Cure Chemistry of Renewable Bis(cyanate) Esters Derived from 2-Methoxy-4-Methylphenol. Biomacromolecules. 2013, 14 (3), 771-780.