2,4-Dimethylthiazole, 99%
2,4-Dimethylthiazole, 99%
2,4-Dimethylthiazole, 99%
2,4-Dimethylthiazole, 99%
Thermo Scientific Chemicals

2,4-Dimethylthiazole, 99%

CAS: 541-58-2 | C5H7NS | 113.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13604.14
also known as A13604-14
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.10
Save 14.45 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.10
Save 14.45 (15%)
Each
Add to cart
Chemical Identifiers
CAS541-58-2
IUPAC Name2,4-dimethyl-1,3-thiazole
Molecular FormulaC5H7NS
InChI KeyOBSLLHNATPQFMJ-UHFFFAOYSA-N
SMILESCC1=NC(C)=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5070-1.5110 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only