2-Hydroxybenzonitrile, 98%
2-Hydroxybenzonitrile, 98%
2-Hydroxybenzonitrile, 98%
Thermo Scientific Chemicals

2-Hydroxybenzonitrile, 98%

CAS: 611-20-1 | C7H5NO | 119.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13606.06
also known as A13606-06
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.60
Save 6.95 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.60
Save 6.95 (15%)
Each
Chemical Identifiers
CAS611-20-1
IUPAC Name2-hydroxybenzonitrile
Molecular FormulaC7H5NO
InChI KeyCHZCERSEMVWNHL-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormCrystalline powder or powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
View more
2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Yuri I. Binev.; Miglena K. Georgieva.; Lalka I. Daskalova. Spectrochemical, ab initio and density functional studies on the conversion of 2-hydroxybenzonitrile (o-cyanophenol) into the oxyanion. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2004, 60, (11), 2601-2610.
  2. Florent Bonnichon.; Claire Richard. Phototransformation of 3-hydroxybenzonitrile in water. Journal of Photochemistry and Photobiology A: Chemistry. 1998, 119, (1), 25-32.