Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate, 97%
Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate, 97%
Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate, 97%

CAS: 155377-19-8 | C7H7F3N2O2 | 208.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13609.14
also known as A13609-14
Price (USD)
264.65
Online Exclusive
294.00
Save 29.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
264.65
Online Exclusive
294.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS155377-19-8
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to pale yellow
Melting Point (clear melt)141.0-147.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only