3-Dimethylaminobenzoic acid, 98%
3-Dimethylaminobenzoic acid, 98%
3-Dimethylaminobenzoic acid, 98%
Thermo Scientific Chemicals

3-Dimethylaminobenzoic acid, 98%

CAS: 99-64-9 | C9H11NO2 | 165.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13612.22
also known as A13612-22
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS99-64-9
IUPAC Name3-(dimethylamino)benzoic acid
Molecular FormulaC9H11NO2
InChI KeyNEGFNJRAUMCZMY-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Appearance (Color)Pale cream to cream to brown
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)147.5-153.5?C
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

Solubility
Soluble in methanol (50 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. W.L. Baker.; Kunamalar Sabapathy.; Maria Vibat.; G. Lonergan. Laccase catalyzes formation of an indamine dye between 3-methyl-2-benzothiazolinone hydrazone and 3-dimethylaminobenzoic acid. Enzyme and Microbial Technology. 1996, 18 (2), 90-94.
  2. Padmarajaiah Nagaraja.; Anantharaman Shivakumar.; Ashwinee Kumar Shrestha. Peroxidase-Catalyzed Oxidative Coupling of Paraphenylenediamine with 3-Dimethylaminobenzoic Acid: Application in Crude Plant Extracts. J. Agric. Food Chem. 2007, 57 (12), 5173-5177.