Flavone, 99%
Flavone, 99%
Flavone, 99%
Thermo Scientific Chemicals

Flavone, 99%

CAS: 525-82-6 | C15H10O2 | 222.243 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13627.06
also known as A13627-06
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
88.50
Save 12.85 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
88.50
Save 12.85 (15%)
Each
Chemical Identifiers
CAS525-82-6
IUPAC Name2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O2
InChI KeyVHBFFQKBGNRLFZ-UHFFFAOYSA-N
SMILESO=C1C=C(OC2=CC=CC=C12)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)95-98?C
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
Flavone is a potentially useful biochemical for cytochrome P450 studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavone is a potentially useful biochemical for cytochrome P450 studies.

Solubility
Soluble in acetone (25 mg/ml), methanol, alcohol, and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers.Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. NK Peters.; JW Frost.; SR Long. A plant flavone, luteolin, induces expression of Rhizobium meliloti nodulation genes. Science. 1986, 233 (4767), 977-980 .
  2. C La Casa.; I Villegas.; C Alarcón de la Lastra.; V Motilva.; M.J Martın Calero. Evidence for protective and antioxidant properties of rutin, a natural flavone, against ethanol induced gastric lesions. Journal of Ethnopharmacology. 2000, 71 (1-2), 45-53.