2-Furoic acid hydrazide, 98%
2-Furoic acid hydrazide, 98%
2-Furoic acid hydrazide, 98%
2-Furoic acid hydrazide, 98%
Thermo Scientific Chemicals

2-Furoic acid hydrazide, 98%

CAS: 3326-71-4 | C5H6N2O2 | 126.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13630.09
also known as A13630-09
Price (USD)
57.65
Online Exclusive
64.40
Save 6.75 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
64.40
Save 6.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS3326-71-4
IUPAC Namefuran-2-carbohydrazide
Molecular FormulaC5H6N2O2
InChI KeySKTSVWWOAIAIKI-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)76.0-82.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only