2-Bromothioanisole, 98%
2-Bromothioanisole, 98%
2-Bromothioanisole, 98%
Thermo Scientific Chemicals

2-Bromothioanisole, 98%

CAS: 19614-16-5 | C7H7BrS | 203.097 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13632.14
also known as A13632-14
Price (USD)
326.65
Online Exclusive
363.00
Save 36.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
326.65
Online Exclusive
363.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS19614-16-5
IUPAC Name1-bromo-2-(methylsulfanyl)benzene
Molecular FormulaC7H7BrS
InChI KeyALAQDUSTXPEHMH-UHFFFAOYSA-N
SMILESCSC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow or pale grey
Refractive Index1.6315-1.6355 @ 20°C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only