5-Bromoisatin, 90+%
5-Bromoisatin, 90+%
5-Bromoisatin, 90+%
Thermo Scientific Chemicals

5-Bromoisatin, 90+%

CAS: 87-48-9 | C8H4BrNO2 | 226.029 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number A13641.18
also known as A13641-18
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.00
Save 12.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.00
Save 12.35
Each
Chemical Identifiers
CAS87-48-9
IUPAC Name5-bromo-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4BrNO2
InChI KeyMBVCESWADCIXJN-UHFFFAOYSA-N
SMILESBrC1=CC=C2NC(=O)C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Assay (HPLC)≥90.0%
Water Content (Karl Fischer Titration)≤5%
Identification (FTIR)Conforms
FormPowder
5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.

Solubility
Soluble in N,N dimethylformamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sumpter WC. The Structures of the Bromodioxindoles of Baeyer and Knop. J. Am. Chem. Soc. 1945, 67 (7), 1140-1141.
  2. Sarkis GY. Synthesis and spectral data for cinchoninic acids. J. Chem. Eng. Data. 1972, 17 (3), 388-391.