Octanophenone, 98%
Octanophenone, 98%
Octanophenone, 98%
Thermo Scientific Chemicals

Octanophenone, 98%

CAS: 1674-37-9 | C14H20O | 204.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13652.06
also known as A13652-06
Price (USD)
47.65
Online Exclusive
53.30
Save 5.65 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
53.30
Save 5.65 (11%)
Each
Chemical Identifiers
CAS1674-37-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to cream
Refractive Index1.5030-1.5070 @ 20?C
FormLiquid
Assay (GC)≥97.5%
Octanophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanophenone is used as a pharmaceutical intermediate.

Solubility
Soluble in hot ethanol, acetone, toluene and other common organic solvents. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Martin Sandvoss.; Ben Bardsley.; Tony L. Beck.; Emma Lee-Smith.; Stephanie E. North.; Peter J. Moore.; Andrew J. Edwards.; Richard J. Smith. HPLC-SPE-NMR in pharmaceutical development: capabilities and applications. Magnetic Resonance in Chemistry. 2005, 43 (9), 762-770.
  2. I. M. Mutton. Use of short columns and high flow rates for rapid gradient reversed-phase chromatography. Chromatographia. 1998, 47 (5), 291-298.