3-(4-Methoxyphenyl)propionic acid, 98%
3-(4-Methoxyphenyl)propionic acid, 98%
3-(4-Methoxyphenyl)propionic acid, 98%
3-(4-Methoxyphenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(4-Methoxyphenyl)propionic acid, 98%

CAS: 1929-29-9 | C10H12O3 | 180.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13655.22
also known as A13655-22
Price (USD)
457.65
Online Exclusive
508.00
Save 50.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
457.65
Online Exclusive
508.00
Save 50.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1929-29-9
IUPAC Name3-(4-methoxyphenyl)propanoic acid
Molecular FormulaC10H12O3
InChI KeyFIUFLISGGHNPSM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CCC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormCrystalline powder or powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (UK-sourced material)
Assay (Silylated GC)≥97.5% (UK-sourced material)
Assay from Supplier's CofA≥97.5% (US-sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only