4-Methoxycoumarin, 98%
4-Methoxycoumarin, 98%
4-Methoxycoumarin, 98%
Thermo Scientific Chemicals

4-Methoxycoumarin, 98%

CAS: 20280-81-3 | C10H8O3 | 176.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13670.06
also known as A13670-06
Price (USD)
70.65
Online Exclusive
78.20
Save 7.55 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS20280-81-3
IUPAC Name4-methoxy-2H-chromen-2-one
Molecular FormulaC10H8O3
InChI KeyMLCMXDYMSAZNPC-UHFFFAOYSA-N
SMILESCOC1=CC(=O)OC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)122.5-128.5?C
Appearance (Color)White
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only