2-Amino-4,6-dimethylpyrimidine, 98%
2-Amino-4,6-dimethylpyrimidine, 98%
2-Amino-4,6-dimethylpyrimidine, 98%
2-Amino-4,6-dimethylpyrimidine, 98%
Thermo Scientific Chemicals

2-Amino-4,6-dimethylpyrimidine, 98%

CAS: 767-15-7 | C6H9N3 | 123.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13674.14
also known as A13674-14
Price (USD)
39.65
Online Exclusive
44.00
Save 4.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.00
Save 4.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS767-15-7
IUPAC Name4,6-dimethylpyrimidin-2-amine
Molecular FormulaC6H9N3
InChI KeyIDQNBVFPZMCDDN-UHFFFAOYSA-N
SMILESCC1=CC(C)=NC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only