Thermo Scientific Chemicals

L-Menthone, 97%, Thermo Scientific Chemicals

Catalog number: A13679.22
100 g, Each
Thermo Scientific Chemicals

L-Menthone, 97%, Thermo Scientific Chemicals

Catalog number: A13679.22
100 g, Each
Quantity
Catalog number: A13679.22
also known as A13679-22
Price (USD)
Price: 106.00
Online price: 90.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14073-97-3
IUPAC Name
(2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one
Molecular Formula
C10H18O
InChI Key
NFLGAXVYCFJBMK-BDAKNGLRSA-N
SMILES
CC(C)[C@@H]1CC[C@@H](C)CC1=O
Appearance (Color)
Clear colorless
Refractive Index
1.4450-1.4550 @ 20?C
Form
Liquid
Optical Rotation
-25? ? 5? (neat)
Assay (GC)
≥96.0%

Description

L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

Solubility
Soluble in ether, alcohol, benzene, acetone.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text