Diethyl iodomethylphosphonate, 98+%
Diethyl iodomethylphosphonate, 98+%
Diethyl iodomethylphosphonate, 98+%
Thermo Scientific Chemicals

Diethyl iodomethylphosphonate, 98+%

CAS: 10419-77-9 | C5H12IO3P | 278.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13697.14
also known as A13697-14
Price (USD)
327.65
Online Exclusive
364.00
Save 36.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
327.65
Online Exclusive
364.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS10419-77-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.4970-1.5010 @ 20?C
Assay (GC)≥98.0%
Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Linden, W. A.; Segal, E.; Child, M. A.; Byzia, A.; Drąg, M.; Bogyo, M. Design and Synthesis of Activity-Based Probes and Inhibitors for Bleomycin Hydrolase. Chem. Biol. 2015, 22 (8), 995-1001.
  2. Li, Y.; Huang, H.; Wang, Z.; Yang, F.; Li, D.; Qin, B.; Ren, X. Stereoselective synthesis of C-sulfonylated aziridines from halomethyl phenyl sulfone and N-tert-butanesulfinyl imines. RSC Adv. 2014, 4 (2), 969-973.