Adipic acid, 99%
Adipic acid, 99%
Adipic acid, 99%
Thermo Scientific Chemicals

Adipic acid, 99%

CAS: 124-04-9 | C6H10O4 | 146.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25,000 g
1000 g
5000 g
Catalog number A13705.0B
also known as A13705-0B
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.50
Save 5.85 (14%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.50
Save 5.85 (14%)
Each
Add to cart
Chemical Identifiers
CAS124-04-9
IUPAC Namehexanedioic acid
Molecular FormulaC6H10O4
InChI KeyWNLRTRBMVRJNCN-UHFFFAOYSA-N
SMILESOC(=O)CCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (U.K. specification)
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.5% (U.K. specification)
CommentPurchased in U.K. and U.S.
Melting Point (clear melt)151-154°C (U.K. specification)
View more
Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.

Solubility
Soluble in methanol and acetone. Slightly soluble in cyclohexane. Insoluble in benzene, acetic acid and petroleum ether.

Notes
Incompatible with oxidizing agents
RUO – Research Use Only

General References:

  1. Vardon, D. R.; Franden, M. A.; Johnson, C. W.; Karp, E. M.; Guarnieri, M. T.; Linger, J. G.; Salm, M. J.; Strathmann, T. J.; Beckham, G. T. Adipic acid production from lignin. Energy Environ. Sci. 2015, 8 (2), 617-628.
  2. Zou, G.; Zhong, W.; Mao, L.; Xu, Q.; Xiao, J.; Yin, D.; Xiao, Z.; Kirk, S. R.; Shu, T. A non-nitric acid method of adipic acid synthesis: organic solvent-and promoter-free oxidation of cyclohexanone with oxygen over hollow-structured Mn/TS-1 catalysts. Green Chem. 2015, 17 (3), 1884-1892.