2-(Phenylthio)thiophene, 97+%
2-(Phenylthio)thiophene, 97+%
2-(Phenylthio)thiophene, 97+%
Thermo Scientific Chemicals

2-(Phenylthio)thiophene, 97+%

CAS: 16718-12-0 | C10H8S2 | 192.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13708.03
also known as A13708-03
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.30
Save 11.65 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.30
Save 11.65 (15%)
Each
Chemical Identifiers
CAS16718-12-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6455-1.6505 @ 20°C
Appearance (Color)Clear pale green to green to brown or pale pink
Assay (GC)≥97.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only