3-Hydroxy-4-methoxybenzoic acid, 97%
3-Hydroxy-4-methoxybenzoic acid, 97%
3-Hydroxy-4-methoxybenzoic acid, 97%
Thermo Scientific Chemicals

3-Hydroxy-4-methoxybenzoic acid, 97%

CAS: 645-08-9 | C8H8O4 | 168.148 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13709.06
also known as A13709-06
Price (USD)
37.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.00
Each
Chemical Identifiers
CAS645-08-9
IUPAC Name3-hydroxy-4-methoxybenzoate
Molecular FormulaC8H7O4
InChI KeyLBKFGYZQBSGRHY-UHFFFAOYSA-M
SMILESCOC1=CC=C(C=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥96.5 to ≤104.5%
Water Content (Karl Fischer Titration)≤0.5%
Assay (Silylated GC)≥96.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only