CTS™ Neurobasal™-A Medium - FAQs
CTS™ Neurobasal™-A Medium - FAQs
View additional product information for CTS™ Neurobasal™-A Medium - FAQs (A1371001)
1 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.