2,3,4,5,6-Pentafluorobenzophenone, 98%
2,3,4,5,6-Pentafluorobenzophenone, 98%
2,3,4,5,6-Pentafluorobenzophenone, 98%
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzophenone, 98%

CAS: 1536-23-8 | C13H5F5O | 272.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13713.06
also known as A13713-06
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS1536-23-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (GC)≥97.5%
Melting Point (clear melt)33.0-40.0?C
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only