2-Fluorophenylhydrazine hydrochloride, 98%
2-Fluorophenylhydrazine hydrochloride, 98%
2-Fluorophenylhydrazine hydrochloride, 98%
2-Fluorophenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

2-Fluorophenylhydrazine hydrochloride, 98%

CAS: 2924-15-4 | C6H8ClFN2 | 162.592 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13716.06
also known as A13716-06
Price (USD)
66.65
Online Exclusive
74.20
Save 7.55 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.20
Save 7.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS2924-15-4
IUPAC Namehydrogen (2-fluorophenyl)hydrazine chloride
Molecular FormulaC6H8ClFN2
InChI KeyVFZYLSYYMHFPSY-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormPowder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only